Order Entry
Puerto Rico
ContactUsLinkComponent
Somso® Animal Cell Model
Somso® Animal Cell Model
Catalog # 470006-482
Supplier:  MARCUS SOMMER
undefined
Somso® Animal Cell Model
Catalog # 470006-482
Supplier:  MARCUS SOMMER
Supplier Number:  ZOS 110/1

Specifications

  • Description:
    Animal Cell
  • Height:
    37 cm
  • Length:
    18 cm
  • No. of keyed items:
    12
  • Cat. No.:
    470006-482
  • Width:
    18 cm
  • Supplier no.:
    ZOS 110/1
  • No. of parts:
    1

Specifications

About this item

High-Quality Model Shows Cell Structures In Exquisite Detail

  • Highly Detailed In Bright Colors For Easy Identification
  • Smaller Size Is Ideal For Individual Lab groups Or Stations
  • 5 Year Manufacturers Warranty

View all the major organelles, including the nucleus, endoplasmic reticulum, mitochondria, ribosomes, Golgi apparatus, and centrioles, enlarged 10,000X, on one three-dimensional, colorful model. The one-piece model is mounted on a height-adjustable stand.