Smilax, Root Slide
470181-278
: Avantor
- Description:Smilax, Root
- Cat. No.:470181-278
- Stain:Quadruple
- Supplier no.:WARD470181-278
- Orientation:Cross section
Greenbrier
- Identifying features clearly distinguishable
- Rigorous quality control standards
Greenbrier. Shows radial arrangement of xylem with heavy endodermis.