ContactUsLinkComponent
Skeletal Muscle, Teased Preparation Slide
Skeletal Muscle, Teased Preparation Slide
Catalog # 470182-672
Supplier:  Avantor
undefined
Skeletal Muscle, Teased Preparation Slide
Catalog # 470182-672
Supplier:  Avantor
Specifications
  • Description:
    Skeletal Muscle, Teased Preparation Slide
  • Cat. No.:
    470182-672
  • Stain:
    Silver impreg.
  • Supplier no.:
    WARD470182-672
  • Orientation:
    Whole mount
Specifications
About this item
Illustrates a voluntary muscle type
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Isolated fibers showing striations and random nuclei.