ContactUsLinkComponent
Lab-Aids® Simulated Urinalysis Kit
Catalog # 470111-834
Supplier:  Lab-Aids
undefined
Lab-Aids® Simulated Urinalysis Kit
Catalog # 470111-834
Supplier:  Lab-Aids
Supplier Number:  4
Specifications
  • Description:
    Simulated Urinalysis Kit
  • No. of lab groups:
    30
  • Cat. No.:
    470111-834
  • Supplier no.:
    4
Specifications
About this item
How do you test urine?
  • Students perform diagnostic lab tests
  • Simulated urine included
  • Time required: one 50 minute class period
Watch any athletic event on TV and the subject of testing for banned substances will always be discussed in some form of fashion. So how do you teach the basics of urinalysis in the classroom? With this kit! Students analyze simulated urine samples to understand the procedures used in urinalysis and the potential applicability of urinalysis as a diagnostic tool. By adding different reagents to urine samples, students observe how each reacts with substances in the urine, providing useful evidence for identifying different substances. Students test simulated urine samples for pH, density, glucose, phosphates and chlorides. Enough reagents are provided for 30 students.
Ordering information: This kit includes a teacher’s guide with SDS, student worksheets and guides, Chemplates with spatula, density tubes, pH color charts, universal Indicator paper and vial, Benedict’s Solution with drop control bottles, chlorine test solution 1 with drop control bottle, phosphate test solution 1 with drop control bottle, Phosphate Test Solution 2 with drop control bottle, Simulated Urine I with drop control bottle, Simulated Urine II with drop control bottle, Simulated Urine III with drop control bottle, Simulated Urine IV with drop control bottle, and Simulated Urine V with Drop control bottle Materials. A hot plate, Bunsen burner, and test tubes are required but not included.
Delivery: See datasheet for contents information.