ContactUsLinkComponent
Lab-Aids® Simulated Blood Typing Kit
Lab-Aids® Simulated Blood Typing Kit
Catalog # 470111-910
Supplier:  Lab-Aids
undefined
Lab-Aids® Simulated Blood Typing Kit
Catalog # 470111-910
Supplier:  Lab-Aids
Supplier Number:  1-S
Specifications
  • Description:
    Simulated Blood Typing Kit
  • No. of lab groups:
    60
  • Cat. No.:
    470111-910
  • Supplier no.:
    1-S
Specifications
About this item
Simulated blood typing for safety.
  • Perform blood typing without human blood
  • Observe agglutination and typing Of A, B, Ab and O blood groups
  • Time required: one 50 minutes class period
Simulated blood typing kit lets students practice the typing of A, B, AB and O blood groups while avoiding the risks associated with real blood. The clumping of simulated blood cells by corresponding simulated antibodies is done through chemical precipitation. Students observe the antigen-antibody relations of artificial bloods and sera. No human animal blood or blood products are used because the reactions are based on chemical solubility.
Included:
LAB-AIDS Slide Guides®
mixing spatulas, blue
mixing spatulas, yellow
bottle simulated Anti-A serum
bottle simulated Anti-B serum
bottle simulated Blood Type A
bottle simulated Blood Type B
bottle simulated Blood Type AB
bottle simulated Blood Type O
Student Worksheet and Guides
Teacher's Guide
SDS (Safety Data Sheet) packet
Delivery: See datasheet for contents information.