Rod and Clamp Assembly
470203-682
- Description:Rod and Clamp Assembly
- Cat. No.:470203-682
- Supplier no.:80780011
This assembly assists with specific gravity determinations.
- Assembly Attaches to Triple Beam Balances
- Two 1000 g Weights Included
- One 500 g Weight Included
- For Making Specific Gravity Determinations
This assembly attaches to a triple beam balance.
It includes three weight sets.
: The attachment weight set includes (2) 1000 g and (1) 500 g weights.
: The attachment weight set includes two 1000 g and one 500 g weights.