ContactUsLinkComponent
Replacement Wicks
Replacement Wicks
Catalog # 470020-700
Supplier:  Avantor
undefined
Replacement Wicks
Catalog # 470020-700
Supplier:  Avantor
Specifications
  • Description:
    Replacement wicks
  • Cat. No.:
    470020-700
  • Supplier no.:
    103-3
Specifications
About this item
Fits Economy and Polyhedral Alcohol Burners.
  • Wick is 7 inches long with a quarter inch diameter
  • Made from woven cotton