Rainbow Fractions® Tiles -n- Work Tray
Catalog # 470046-138
Supplier: Learning Resources
Specifications
- Description:Rainbow Fractions® Tiles -n- Work Tray
- Environmentally Preferable:
- Cat. No.:470046-138
- Supplier no.:LER0615
Specifications
About this item
Pieces of 2.5mm thick solid plastic have fractional values are printed on each tile.
- Learn fractions
- Values printed on each tile
- Includes 51 pieces
The 8" square work tray doubles as a handy storage container. Includes 51 pieces and an activity guide.
Caution: WARNING: CHOKING HAZARD- Small Parts. Not for Children Under 3 Years.