VWR® Qualitative Grade Filter Paper Circles
Supplier: Cytiva
Certificates
About this item
Grade #415.
Grade #410
Porosity: Fine
Flow Rate: Slow
Particle Retention: 1 μm
Applications: Analytical operations involving filtration of CuCO2, PbSO4, BaSO4, MnCO3, etc.