ContactUsLinkComponent
PYREX® Liebig Condenser
PYREX® Liebig Condenser
  470211-398
 :  Corning
undefined
PYREX® Liebig Condenser
  470211-398
 :  Corning
 :  2340-200
  • Description:
    Pyrex® Liebig Condenser, Jacket: 41 mm x 200 mm, Overall Length: 345 mm
  • Cat. No.:
    470211-398
  • Supplier no.:
    2340-200
A fine borosilicate condenser with the inner tube sealed in.
The large-capacity water jacket ensures efficient cooling. The adapter end will accept a No. 3 rubber stopper.