PYREX® Flat-Bottom Evaporating Dish
Pyrex® Flat-Bottom Evaporating Dish
Supplier: Corning
Supplier #: 3180-80
Jump to:
Product Details & Documents
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Dishes with rugged beaded rims and standard medium depth.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Pyrex® Flat-Bottom Evaporating Dish