ContactUsLinkComponent
Prokaryote Survey Slide Set
Prokaryote Survey Slide Set
Catalog # 470177-698
Supplier:  Avantor
undefined
Prokaryote Survey Slide Set
Catalog # 470177-698
Supplier:  Avantor
Specifications
  • Type:
    Monera
  • Description:
    Prokaryote Survey Slide Set
  • No. of slides:
    12
  • Cat. No.:
    470177-698
  • Supplier no.:
    WARD470177-698
Specifications
About this item
An Ideal Guide to the Major Prokaryote Groups
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
All are slides are prepared as smears unless otherwise specified.
Ordering information: Set includes Archaebacteria: Mixed forms; Eubacteria: Typical bacillus, Typical gram + cocci, Typical gram – cocci, Typical spirillum, Cocci in clumps (Staphylococci), Cocci in chains (Streptococci), Bacillus megaterium, Micrococcus luteus, Treponema sp., Root nodule-legume bacteria (sect); Cyanobacteria: Anabaena (wm), and Anacystis (wm).