Polypropylene Powder Funnels
Catalog # 470191-320
Supplier: VWR International
Specifications
- Description:Powder Funnel, Polypropylene, 80 mm diameter, Scienceware
- Cat. No.:470191-320
- Supplier no.:650020
Specifications
About this item
These unbreakable polypropylene powder funnels are ideal for student use. External ribs prevent air lock. Can be autoclaved. Stem length is 35 mm.