Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium Phosphate Monobasic
Potassium Phosphate Monobasic
Catalog # 470302-248
Supplier:  Ward's Science
CAS Number:  
undefined
Potassium Phosphate Monobasic
Catalog # 470302-248
Supplier:  Ward's Science
Supplier Number:  PP0730-100G
CAS Number:  

Specifications

  • Description:
    Potassium Phosphate Monobasic
  • Size:
    100 g
  • Chemical/physical specifications:
    Crystal
  • Cat. No.:
    470302-248
  • Grade:
    Laboratory
  • Supplier no.:
    PP0730-100G

Specifications

About this item

CAS Number: 7778-77-0
Formula Weight: 136.09
Formula: KH2PO4
Density (g/mL): 2.338
Freezing Point (°C): 253
Solubility: Water
Synonyms: Potassium Dihydrogen Phosphate
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.