ContactUsLinkComponent
Polarized Film Slides
Polarized Film Slides
Catalog # 470149-018
undefined
Polarized Film Slides
Catalog # 470149-018
Supplier Number:  P30695/3
Specifications
  • Description:
    Polarized film slides, pair
  • Dimensions:
    5×5 cm (2×2")
  • Environmentally Preferable:
  • Cat. No.:
    470149-018
  • Supplier no.:
    P30695/3
Specifications
About this item
Learn how sunglasses work!
  • Pair of identical slides with polarized film
When this pair of polaroid film squares are aligned, light passes through them appearing much as it would if viewed through sunglasses. However, if one square is rotated 90 degrees, no light passes through at all! A great way to demonstrate the principles of polarization and the wave properties of light.