ContactUsLinkComponent
Supertek® Plastic Hall’s Carriage
Supertek® Plastic Hall’s Carriage
Catalog # 470148-700
undefined
Supertek® Plastic Hall’s Carriage
Catalog # 470148-700
Supplier Number:  P10507
Specifications
  • Description:
    Plastic Hall’s Carriage
  • Cat. No.:
    470148-700
  • Supplier no.:
    P10507
Specifications
About this item
Watch work and energy intercept.
This lightweight, nearly frictionless Hall’s Carriage is perfect for studying the relationship of work and energy. The wide plastic body can carry bulk loads. Oil-free sleeve bearings never need adjustment. Use with our Picket Fence for determining the coefficient of rolling friction. Instructions included. Size: 13 cm L.