Ward'sⓇ Plant Cell Structure and Animal Cell Structure Slide Sets
Catalog # 470183-468
Supplier: Avantor
Specifications
- Description:Plant cell structure set
- No. of slides:7
- Cat. No.:470183-468
- Supplier no.:WARD470183-468
Specifications
About this item
View real examples of plant and animal cells
- Identifying features clearly distinguishable
- Rigorous quality control standards
Our cell structure slides are great additions to your curriculum to teach structure and function or compare and contrast animal and plant cell components to each other.
Plant cell structure set includes: Entire cell; Cystoliths; Chloroplasts; Idioblasts; Mitochondria; Plasmodesmata; Sclerids
Animal cell structure set includes: Entire cell; Mitochondria; Nissl bodies; Centrioles; Golgi apparatus; Glycogen; Lipids
(Animal Cell Structure Set not available in Canada)