Pipette Rack
Catalog # 470144-616
Supplier: United Scientific Supplies
Specifications
- Description:Rack, Pipet
- Cat. No.:470144-616
- Supplier no.:79101
Specifications
About this item
Hold your pipettes securely and within easy reach.
- Three polypropylene coated metal rods
- Autoclavable
Organize the pipettes you use frequently. Holds up to twelve pipettes, six on each side. Polypropylene rack is easy to assemble and quickly disassembles for space saving storage. Inexpensive enough to have one for every lab table. Autoclavable.