About this item
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
- Range: 3 to 10 pH
- Materials for 50 tests
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.