pH Indicator Papers
pH Indicator Papers, Litmus Red, Laboratory, Strips, Strips, Laboratory
Catalog # 470220-896
Supplier: Precision Laboratories
Supplier #: 190-12V-100
Jump to:
Product Details & Documents
pH Test Papers in your Choice of Two Indicator Colors.
- Easy to read
- Vial of 100
Litmus red paper changes to blue at pH 8.1. Color comparison chart not included.
Specifications
- Description:pH Indicator Papers, Litmus Red, Laboratory, Strips
- Grade:Laboratory
- Form:Strips
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



