Penicillium Slide
Catalog # 470181-068
Supplier: Avantor
Specifications
- Description:Penicillium Slide
- Cat. No.:470181-068
- Stain:Quadruple
- Supplier no.:WARD470181-068
- Orientation:Section
Specifications
About this item
Mold structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Orange skin showing mycelia, conidiophores, and conidia.