ContactUsLinkComponent
PCR Based Alu Human DNA Typing
PCR Based Alu Human DNA Typing
Catalog # 470126-560
Supplier:  Edvotek
undefined
PCR Based Alu Human DNA Typing
Catalog # 470126-560
Supplier:  Edvotek
Supplier Number:  333-B
Specifications
  • Description:
    PCR Based Alu Human DNA Typing
  • No. of lab groups:
    5
  • Cat. No.:
    470126-560
  • Supplier no.:
    333-B
Specifications
About this item
An Interactive DNA Investigation
  • Investigate your own DNA
  • Compare your class results to the world's
  • Time required: ~75 min plus 2 hours for PCR
Working with primers for a 300 base pair Alu insertion in chromosome 16 (PV92) students will determine their own unique genotype in this hands-on investigation. Then, compare class results over the Internet with others around the world.
Kit includes: instructions, proteinase K, PCR EdvoBeads™, control DNA and primers, microtubes, agarose, DNA ladder, practice gel loading solution, gel loading dye, buffer, InstaStain® Ethidium Bromide and FlashBlue™ stain.
Required, but not included: micropipettes to measure between 5 and 50 µl, tips, water bath, thermal cycler, electrophoresis apparatus, power supply, microwave or hot plate, UV transilluminator.
Packaging: Some components require freezer storage on arrival