Specifications
- Description:Patch Cord, Banana/Alligator, 24"
- Cat. No.:470020-588
- Supplier no.:P56522
Specifications
About this item
Flexible, 18 gauge wire with a banana plug at one end and an alligator clip at the other.
The alligator clip has a protective vinyl sheath.