ContactUsLinkComponent
Lab-Aids® Natural Selection And Antibiotic-Resistant Bacteria Kit
Lab-Aids® Natural Selection And Antibiotic-Resistant Bacteria Kit
  470024-334
 :  Lab-Aids
undefined
Lab-Aids® Natural Selection And Antibiotic-Resistant Bacteria Kit
  470024-334
 :  Lab-Aids
 :  904S
  • Description:
    Antibiotic-Resistant Bacteria Kit
  • No. of lab groups:
    30
  • Cat. No.:
    470024-334
  • Supplier no.:
    904S
Model The Effects Of Antibiotics On Disease Causing Bacteria During Infection
  • Demonstrate The Importance Of Taking Antibiotics As Prescribed
  • Re-Create The Interactions Of Antibiotics With Bacteria Populations
  • Time Required: One To Two 50 Minute Class Periods
Demonstrate the importance of taking antibiotic prescriptions as prescribed with this simulation kit. Students re-create the interactions of antibiotics with bacteria populations, with some groups completing the entire course, while other groups miss doses. The populations of more and less-resistant bacteria are then graphed for comparison, and students observe the selection of more-resistant bacteria that can take place if antibiotic treatment is intermittent or discontinued.
Included:
Teacher’s Guide with SDS
Student Worksheets and Guides
Blue disks
Yellow disks
Red disks
Number cubes
Transparency: Bacteria Graph Sample 1
Transparency: Bacteria Graph Sample2
Bacteria Graphs
 : See datasheet for contents information.