ContactUsLinkComponent
Munsell® Soil Color Chart
Munsell® Soil Color Chart
Catalog # 470003-876
Supplier:  Peco Sales
undefined
Munsell® Soil Color Chart
Catalog # 470003-876
Supplier:  Peco Sales
Supplier Number:  77321
Specifications
  • Description:
    Munsell® Soil Color Chart
  • Environmentally Preferable:
  • Cat. No.:
    470003-876
  • Supplier no.:
    77321
Specifications
About this item
Features Over 300 Color Chips
Approximately 322 chips are permanently mounted on charts in a loose-leaf binder to make it easy to compare soil color, rocks, and archaeological specimens. The chart also includes two Gley Charts for easier comparison. Size: 7 5/8" x 5 5/8".