Motor Generator
Generator, Motor
Catalog # 470005-672
Supplier: Wards
: PH1241
Jump to:
Product Details & Documents
An excellent apparatus for showing the function of parts in an AC or DC motor and generator. Operates as either a shunt-wound or series motor. The unit is similar to the St. Louis Motor. Included with the motor are a powerful horseshoe magnet and an electromagnet.
Specifications
- Description:Generator, Motor