ContactUsLinkComponent
Mohr’s Pinchcock Tubing Clamp
Mohr’s Pinchcock Tubing Clamp
  470104-106
Mohr’s Pinchcock Tubing Clamp
  470104-106
 :  H-8610
  • Description:
    Clamp, Tubing, 16 mm Tubing, Mohr's Pinchcock (5/8") OD, Humboldt
  • Environmentally Preferable:
  • Cat. No.:
    470104-106
  • Supplier no.:
    H-8610
This clamp is made from one-piece, rectangular, nickel-plated brass spring wire with rounded edges.
It is self aligning and has molded finger grips.