ContactUsLinkComponent
Midget Urinometer, Jar Only
Midget Urinometer, Jar Only
  470148-868
undefined
Midget Urinometer, Jar Only
  470148-868
 :  MIDGETJAR
  • Description:
    Midget Urinometer, Jar Only
  • Cat. No.:
    470148-868
  • Supplier no.:
    MIDGETJAR
For highly accurate samples when only minimal volumes are required, use this simple, stain-free apparatus.
Made of high-quality clear glass, this 50 ml urinometer has a wide bottom to prevent tipping and a non-stick float. It includes instructions. Range: 1.000 to 1.060 Sp. Gr in 0.002 graduations.