ContactUsLinkComponent
Metabolism Experiment Kit
Metabolism Experiment Kit
Catalog # 470002-468
Supplier:  Lab-Aids
undefined
Metabolism Experiment Kit
Catalog # 470002-468
Supplier:  Lab-Aids
Supplier Number:  55
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Metabolism Experiment Kit
  • Environmentally Preferable:
  • No. of lab groups:
    10
  • Cat. No.:
    470002-468
  • Supplier no.:
    55
Specifications
About this item
With the Metabolism Experiment Kit, students draw correlations between organism size, oxygen consumption, and temperature.
  • Time Required: One 50 Minute Class Period
  • Excellent Introduction to Metabolic Process
  • Teacher’s Guide with SDS Provided
Students measure the rate of oxygen consumption of small organisms such as insects, snails, worms, and germinating seeds. Using Patented (U.S. Pat. No. 3,581,737) LAB-AIDS® Metabolism chambers, they observe the organisms and draw correlations between the size of an organism and the temperature of the environment and oxygen consumption. They also calculate an organism’s respiratory rate in cubic cm of oxygen per hour, measure total gas change in a closed environment, and determine an organism’s metabolic relationship to total gas in a closed environment.
The kit includes two unbreakable chambers large enough for small animals and plants. One chamber is used to measure the organism's oxygen consumption, and the other serves as a control.
Ordering information: This kit includes metabolism chambers, carbon dioxide absorbent, capillary tubes with a connecting tube (calibrated), forceps, latticed organism holders with plastic retainers, index solution (colored, vial), student data cards, and a teacher’s guide with SDS. Animal or plant specimens for testing are needed but not included.