ContactUsLinkComponent
Lily Meiosis, First Meiotic Division Slide
Lily Meiosis, First Meiotic Division Slide
Catalog # 470181-254
Supplier:  Avantor
undefined
Lily Meiosis, First Meiotic Division Slide
Catalog # 470181-254
Supplier:  Avantor
Specifications
  • Description:
    Lily Meiosis, First Meiotic Division Slide
  • Cat. No.:
    470181-254
  • Stain:
    Quadruple
  • Supplier no.:
    WARD470181-254
  • Orientation:
    Cross section
Specifications
About this item
Investigate flowering plant meiosis
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows metaphase of first meiotic division.