ContactUsLinkComponent
Laboratory Safety Mat
Laboratory Safety Mat
Catalog # 470016-916
Supplier:  Fischer Technical
undefined
Laboratory Safety Mat
Catalog # 470016-916
Supplier:  Fischer Technical
Supplier Number:  MATRED
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Red
  • Description:
    Laboratory Safety Mat
  • Thickness:
    ⅛"
  • Cat. No.:
    470016-916
  • W×L:
    9 1/2 × 9 1/2"
  • Supplier no.:
    MATRED
Specifications
About this item
Stabilize glassware on orbital or rocking shakers and eliminate slippery conditions on benchtop surfaces with this versatile non-slip safety mat.
Its ribbed, textured surface steadily holds beakers, cylinders, and other glassware in place, even at an angle. A valuable addition to any laboratory, the silicone rubber mat can be autoclaved, cleaned with disinfectants such as bleach, easily cut to various sizes, and will withstand temperatures up to 260°C.
Size: 9 1/2" square x 1/8" thick.