pH Measurement/Indicator Kit
About this item
Students explore the wide range of acid and bases and their effect on indicator dyes with this kit.
- Activities Demonstrate pH's Relationship to Acids, Bases, and Indicator Dyes
- Students Create Individual pH Color Guides
- Colorful Tests Develop a Better Understanding of pH
- Color Guides Identify pH of Three Unknowns
- Required Materials Provided
In this popular Applied Science Concept Kit, students perform a series of colorful tests that help them develop a better understanding of pH and its relationship to acids and bases.
Using LAB-AIDS Chemplates® and a universal indicator, students creates their own individual pH color guide. They use this color guide to identify the pH of the three unknowns, which are provided in this kit.
Ordering information: This kit includes Chemplates, LAB-AIDS, bromothymol blue, methyl orange, phenolphthalein, a universal indicator, pH 1; 3; 5; 7; 9; 11; and 13, and unknowns #1; #2; and #3.
1 Options Available Below
Product Details & Documents
Students explore the wide range of acid and bases and their effect on indicator dyes with this kit.
- Activities Demonstrate pH's Relationship to Acids, Bases, and Indicator Dyes
- Students Create Individual pH Color Guides
- Colorful Tests Develop a Better Understanding of pH
- Color Guides Identify pH of Three Unknowns
- Required Materials Provided
In this popular Applied Science Concept Kit, students perform a series of colorful tests that help them develop a better understanding of pH and its relationship to acids and bases.
Using LAB-AIDS Chemplates® and a universal indicator, students creates their own individual pH color guide. They use this color guide to identify the pH of the three unknowns, which are provided in this kit.
Ordering information: This kit includes Chemplates, LAB-AIDS, bromothymol blue, methyl orange, phenolphthalein, a universal indicator, pH 1; 3; 5; 7; 9; 11; and 13, and unknowns #1; #2; and #3.
Product Family Options
Product Information
- IncludesNo. of lab groupsAvailabilityPrice
- 470105-620pH Measurement and Indicators Refill KitSupplier Number: 81R-RC
- Discontinued-Supplier Number: -
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



