Kidney, Mammal Slide
Catalog # 470182-926
Supplier: Avantor
Specifications
- Description:Kidney, Mammal
- Cat. No.:470182-926
- Stain:Periodic acid Schiff reaction
- Supplier no.:WARD470182-926
- Orientation:Section
Specifications
About this item
Illustrates Structures of the Kidney
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Stained for basement membrane of tubules, Bowman’s capsules, and brush borders of proximal tubules.