ContactUsLinkComponent
Iron-Deficiency Anemia Slide
Iron-Deficiency Anemia Slide
Catalog # 470177-660
Supplier:  Avantor
undefined
Iron-Deficiency Anemia Slide
Catalog # 470177-660
Supplier:  Avantor
Specifications
  • Description:
    Iron-Deficiency Anemia Slide
  • Environmentally Preferable:
  • Cat. No.:
    470177-660
  • Stain:
    Giemsa
  • Supplier no.:
    WARD470177-660
  • Orientation:
    Smear
Specifications
About this item
Illustrates red blood cell characteristics of the deficiency
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Peripheral blood. Characterized by small, pale red blood cells.