Human Skin, Composite Slide
Human Skin, Composite Slide, Section, Hematoxylin and Eosin
Catalog # 470183-070
Supplier: Avantor
Jump to:
Product Details & Documents
Study human epidermis
- Identifying features clearly distinguishable
- Rigorous quality control standards
Pigmented and unpigmented adult skin with all layers, epidermis, dermis, and subcutaneous tissue, for easy comparison.
Specifications
- Description:Human Skin, Composite Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section