Lab-Aids® Human Genetics Experiment Kit
Catalog # 470111-670
Supplier: Lab-Aids
Specifications
- Description:Human Genetics Experiment Kit
- Environmentally Preferable:
- No. of lab groups:50
- Cat. No.:470111-670
- Supplier no.:7
Specifications
About this item
How are inherited traits transmitted?
- Develop an experiment
- Understand and demonstrate principles of heredity
- Time required: one 50 minutes class period
The experiment is based on the transmission of a single inherited trait - the ability to taste harmless PTC (phenylthiocarbamide). The goal is for students to determine if they have this gene. They collect data by testing family members, correlating and pooling the data for the entire class, then calculating the random segregation data according to the Hardy-Weinberg Law. Background information about the Hardy-Weinberg Law and information about calculating gene frequency and the genetics of ABO blood typing are included.
Included:
Teacher's Guide with SDS
PTC test paper, strips
Control test paper, strips
Genealogy and data charts
Delivery: See datasheet for contents information.