Specifications
- Description:H-racer 2.0 IR Control
- Height:3.9"
- Length:11.8"
- No. of lab groups:1
- Cat. No.:470145-606
- Width:8.15"
- Supplier no.:FCJJ-23-B
Specifications
About this item
Build Your Own Fuel Cell Car!
- Award winning car design
- Components are visible during operation
- 12 years old and up
Experience the automotive technology of the future by building and driving your own hydrogen fuel cell car. The latest generation of the H-racer, H-Racer 2.0, allows you to watch the process of electrolysis and the conversion of hydrogen into electrical energy— Both the car and the hydrogen refueling station have transparent casing and LED lighting to showcase the science. This design has garnered numerous awards, including: Time Magazine’s “Best Inventions”, Business Week IDEA, Metropolitan Home “Design 100″, ID magazine annual review, and Business 2.0 Bottom Line Design Awards.
Restriction Comments:
This kit is intended only for use by person 12 years old and up, and only under the supervision of an adult who has read and understood the instructions provided in the kit’s user manual.
WARNING: CHOKING HAZARD–Children under 8 yrs. can choke or suffocate on uninflated or broken balloons. Adult supervision required. Keep uninflated balloons from children. Discard broken balloons at once.
• The balloon is made of natural rubber latex. Children under 12 can choke or suffocate on uninflated or broken balloons. Keep the balloon away from children. Discard a broken balloon at once.
• The Hydrogen Station generates gases that are easily ignited. Do not use the Hydrogen Station for any other purpose than refueling the H-racer, as instructed in the user manual. Keep the Hydrogen Station and H-racer away from flames or fires.