Gymnosperm and Angiosperm Stem Combination Slide
470181-442
: Avantor
- Description:Gymnosperm & Angiosperm Stem Combination
- Cat. No.:470181-442
- Stain:Quadruple
- Supplier no.:WARD470181-442
- Orientation:Cross section
Compare and Contrast Tree Types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Pinus and Tilia. Comparison of hard and soft wood.