Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Guaiacol

Guaiacol

Guaiacol, Laboratory, 250 mL
Catalog # 470301-192
Supplier:   Ward's Science
Supplier #:  GG0231-250ML
Guaiacol
Catalog # 470301-192
Supplier:  Ward's Science
CAS Number:  
Guaiacol
Guaiacol

Guaiacol, Laboratory, 250 mL

CAS Number:  
Price
$104.70
Log in for availability
Each (250ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 90-05-1
Formula Weight: 124.14
Formula: C7H8O2
Density (g/mL): 1.120
Boiling Point (°C): 205
Freezing Point (°C): 28
Solubility: Water
Synonyms: 2-Methoxyphenol
Shelf Life (months): 36
Storage: Red

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Tip: If your Guaiacol has solidied at room temperature, place your bottle in hot tap water for several minutes.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Documents

Specifications

Specifications

  • Description:
    Guaiacol
  • Size:
    250 mL
  • Chemical/physical specifications:
    Solution
  • Grade:
    Laboratory

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.