- Description:Fuel Cell Car Science Kit
- Height:5.5"
- Length:12.6"
- No. of lab groups:1
- Cat. No.:470003-572
- Width:9.25"
- Supplier no.:FCJJ-11
Use Clean Energy to Power a Car that You Build!
- Components are visible during operation
- Assembled car can steer itself
- Includes an assembly manual
Experience the excitement of new fuel cell technology with this customized construction kit. In building this car, you will discover the possibility of using 100% clean fuel to power cars of the future. Once assembled, the car can steer itself independently and will reverse itself when it hits a barrier. A fully functioning vehicle, the car in this kit demonstrates the power of renewable energy on a small, user-friendly scale. A manual includes step by step instructions for simple assembly. All major car components are included - a fuel cell, a car chassis, a small electric motor, two water and gas storage containers, cables, tubes, a syringe, and more. All you need to add is water and two AA batteries.
WARNING: CHOKING HAZARD - Small parts. Not for children under 3 yrs.