Orders LinkContactUsLinkComponent
Faraday Cage
Faraday Cage
Faraday Cage
Catalog # 470039-428
Supplier:   Science First
Supplier #:  615-3095
Faraday Cage
Catalog # 470039-428
Supplier:  Science First
Faraday Cage
Faraday Cage
Faraday Cage
Price
$45.45
Log in for availability
Each
Product Details & Documents
Demonstrate that a charge cannot exist inside a conductor and re-create Faraday’s classic 'Ice pail' experiment with this kit.
  • With instructions
  • Ø×H: 12×27 cm
Delivery information: Includes a perforated metal cage with stand and cover as well as a mounted metal point for the lightning rod effect.
Specifications
  • Description:
    Faraday Cage