ContactUsLinkComponent
Extension Buret Clamp
Extension Buret Clamp
Catalog # 470202-864
undefined
Extension Buret Clamp
Catalog # 470202-864
Supplier Number:  H-8562
Specifications
  • Description:
    Clamp, Buret, Extension, w/ Plastic Coated Round Jaws, Humboldt
  • Cat. No.:
    470202-864
  • Supplier no.:
    H-8562
Specifications
About this item
The plastic-coated symmetrical round jaws on this affordable clamp are ideal for holding objects 9.5-35 mm in diameter.
The overall length of the clamp is 20 cm. Similar to 61105-02.