Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Electric Timer

Electric Timer

Electric timer
Catalog # 470202-492
Supplier:   DIMCOGRAY COMPANY
Supplier #:  7-171-160R
Electric Timer
Catalog # 470202-492
Supplier:  DIMCOGRAY COMPANY
Electric Timer
Electric Timer

Electric timer

Price
$514.85
Log in for availability
Each
Product Details & Documents

This sensitive lab instrument is accurate to ±0.15% at its maximum setting.

  • Maximum load: 1200 W, ¹/₃ hp
  • Range: 60 minutes (minutes/seconds)
  • Dial size: 6¹/₂"Ø

The dial’s larger numbers show elapsed time, while the smaller numbers can be used as a stop clock. It is composed of a sturdy polycarbonate case, built-in buzzer, and two separate three-wire outlet receptacles for automatic switching of appliances.

Specifications
  • Description:
    Electric timer
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.