ContactUsLinkComponent
Demonstration, Eddy Current Experiment
Demonstration, Eddy Current Experiment
Catalog # 470003-662
undefined
Demonstration, Eddy Current Experiment
Catalog # 470003-662
Supplier Number:  EDYCR1
Specifications
  • Description:
    Demonstration, Eddy Current Experiment
  • Environmentally Preferable:
  • Cat. No.:
    470003-662
  • Supplier no.:
    EDYCR1
Specifications
About this item
The Eddy Current Experiment demonstrates Lenz's Law.
  • Copper Tube Features End Caps for Self-Contained Storage
  • Copper Tube Length: 33 cm
  • Neodymium Creates Electrical Current that Resists Falling
  • Experiment Demonstrates Lenz's Law
Two test masses are sent through a 33 cm copper tube. One mass drops quickly, but the other seems to defy gravity by slowing as it falls.
It does this because one mass is steel, and the other is neodymium, a magnet that creates a moving magnetic field, which produces an electrical (eddy) current that resists its fall.