ContactUsLinkComponent
Dual Output Electrophoresis Power Supply
Dual Output Electrophoresis Power Supply
Catalog # 470130-692
Supplier:  VWR International
undefined
Dual Output Electrophoresis Power Supply
Catalog # 470130-692
Supplier:  VWR International
Specifications
  • Description:
    Dual Output Electrophoresis Power Supply
  • Environmentally Preferable:
  • Cat. No.:
    470130-692
  • Supplier no.:
    PH 1341
Specifications
About this item
Double the Voltage
Featuring two different voltages, 70 V or 125 V, this dual output power supply is flexible. The output voltage is easily selected via a rocker switch and is available on two parallel pairs of safety sockets. The unit’s size is 7 × 5.5 × 4" (18 × 14 × 10 cm).