Lab-Aids® DNA, RNA Protein Synthesis Kit
Catalog # 470111-912
Supplier: Lab-Aids
Specifications
- Description:DNA , RNA Protein Synthesis Kit
- No. of lab groups:12
- Cat. No.:470111-912
- Supplier no.:72
Specifications
About this item
This kit demystifies DNA's and RNA's connection to protein synthesis.
- Demystify the DNA/RNA connection
- Model replication, transcription, and translation
- Time required: one 50 minutes class period
Students construct a model of DNA, replicate it, and use half of the ladder to build messenger RNA (mRNA). Placing mRNA on a model ribosome simulates transcription in the nucleus and the movement of mRNA. Students simulate translation by matching the mRNA and its codons to transfer RNA (tRNA) and its anti-codons.
Amino acids bond to form a protein molecule, and students experience how information stored in DNA is expressed as a finished protein.
Delivery: See datasheet for contents information.