Order Entry
Puerto Rico
ContactUsLinkComponent
Digital pH Meter Kit with RS-232 Output
Digital pH Meter Kit with RS-232 Output
Catalog # 470019-736
undefined
Digital pH Meter Kit with RS-232 Output
Catalog # 470019-736
Supplier Number:  DPH7011
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Digital pH Meter Kit with RS-232 Output
  • W×D×H:
    1.6×1.4×7.7"
  • pH Range:
    −2...16
  • Cat. No.:
    470019-736
  • Supplier no.:
    DPH7011

Specifications

About this item

Lightweight, Versatile Unit

  • Measures multiple conditions
  • Durable Housing
  • Easy to Read

The digital display of this pH meter makes readings easy to view and record. Device features automatic temperature compensation, minimum/maximum value recall, and data hold capabilities. Pocket-sized for portability into the field, the waterproof unit is designed to float if dropped into water. Includes soaking solution, pH 4 and 7 buffer solutions, four AAA batteries, and a carrying case. pH range: -2 to 16. ORP range: -1000 to 1000. Temperature range: 32° to 194°F (0° to 90°C). Size: 7.7"L x 1.6"W x 1.4"H. Weight: 4.8 oz.

pH Range: 0 - 14. Resolution: 0.01pH. Accuracy: ±0.01pH.
ORP Range: -1000 to +1000mV. Resolution: 1mV. Accuracy: 2mV.
Temperature Range: 32 - 194°F; 0 - 90° C. Resolution: 0.1°F/°C. Accuracy: 0.4°F/0.2°C.