Orders LinkContactUsLinkComponent
Decade Inductance Box
Decade Inductance Box
Decade Inducatance Box
Catalog # 470121-490
Supplier:   Wards
Supplier #:  PH1135F
Decade Inductance Box
Catalog # 470121-490
Supplier:  Wards
Decade Inductance Box
Decade Inductance Box
Decade Inducatance Box
Clearance
$179.25
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
Investigating Inductance has never been this easy
This sturdy decade box allows a range of inductances from 10 H down to 1 microH. Maximum capacity is 11.11111 H. Housed in a box with seven dials and fitted with 4 mm output plugs, the unit is fully equipped for laboratory use.
Specifications
  • Description:
    Decade Inducatance Box