Day’s Pinchcock Tubing Clamp
Catalog # 470104-108
Supplier: Humboldt Manufacturing
Specifications
- Description:Day’s Pinchcock Tubing Clamp
- Cat. No.:470104-108
- Supplier no.:H-8630
Specifications
About this item
A simple wire clamp that may be installed or removed without disconnecting tubing ends.
Made from nickel-plated steel wire with an overall length of 66 mm (2 5/8"). For tubing up to 9.5 mm O.D.