ContactUsLinkComponent
Drosophilia Cryolizer-Anesthetizer
Drosophilia Cryolizer-Anesthetizer
Catalog # 470002-086
Supplier:  Kemtec
undefined
Drosophilia Cryolizer-Anesthetizer
Catalog # 470002-086
Supplier:  Kemtec
Supplier Number:  2-800
Specifications
  • Description:
    Cryolizer-anesthetizer, Drosophilia
  • Cat. No.:
    470002-086
  • Supplier no.:
    2-800
Specifications
About this item
This plastic easy-to-use cooling unit immobilizes flies for over thirty minutes.
  • Aanesthetizes without sterilizing or killing specimens
  • With Petri dish: Placed in a freezer, it cools flies to immobilize them
Dia. 3½"; height 3½"